Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, DILUT-IT™ Analytical Concentrate, 1N, 1 equiv. = 36.46g HCl, J.T. Baker™

Catalog No. 02003021 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 Pk.
Packaging:
Poly Ampoule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-021 1 Pk. Poly Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-021 Supplier Avantor Supplier No. 465701
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Analytical concentrate for the preparation of a 1 N hydrochloric acid solution.

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Packaging Poly Ampoule
Specific Gravity 1L = 1.06kg
Quantity 1 Pk.
Grade Not Available
Chemical Name or Material Hydrochloric Acid, DILUT-IT™ Analytical Concentrate, 1N, 1 equiv. = 36.46g HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.