Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, Omnipure™ MilliporeSigma™

Catalog No. MHX06071 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MHX06071 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MHX06071 Supplier MilliporeSigma Supplier No. HX06071
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrochloric acid is an economical choice for sensitive trace metal analysis and sample preparation.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Melting Point -74°C
Boiling Point 110°C
Packaging Poly Coated Glass Bottle
Assay Percent Range 34 to 37%
Quantity 500 mL
Linear Formula HCl in water
Formula Weight 36.46
Percent Purity 34% to 37%
Grade Not Available
Chemical Name or Material Hydrochloric Acid, OmniPure™
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.