Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, FCC, 37%, Spectrum™ Chemical
SDP

Catalog No. 18603186 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 L
4 L
20 L
4 x 2.5 L
200 L
Packaging:
Drum
Glass Bottle
Poly Drum
Poly-Coated Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-186 200 L Drum
18-603-184 2.5 L Glass Bottle
18-603-181 4 L Poly-Coated Glass Bottle
18-603-185 4 x 2.5 L Glass Bottle
18-603-183 20 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-603-186 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HY106200LTBL

missing translation for 'validMainframeMsg'

HY106, 7647-01-0, HCl

Spectrum™ Chemical Hydrochloric Acid, 37 Percent, FCC is used in food processing, or as a food additive to adjust the pH. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

CAS Max % 62 to 64%, 36 to 38%
Non_volatile Matter 0.5%
Packaging Drum
Reducing Substances 0.007%
Quantity 200 L
UN Number UN1789
Percent Purity 36 to 38%
Grade FCC
Chemical Name or Material Hydrochloric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.