Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid fuming, 37.0%, for analysis EMSURE™ ACS, ISO, Reag. Ph Eur, MilliporeSigma™

Catalog No. M1003171000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1003171000 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1003171000 Supplier MilliporeSigma Supplier No. 1.00317.1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity ACS reagent grade hydrochloric acid suitable for analytical and regulated laboratory applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym Hydrogen chloride solution
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH < 1 (H2O, 20°C)
Density 1.19 g/cm3 (at 20°C)
Packaging Glass Bottle
Vapor Pressure 190 hPa (20°C)
Assay Percent Range 37.0 to 38.0%
Quantity 1 L
Percent Purity 37.0 to 38.0%
Grade Not Available
Chemical Name or Material Hydrochloric Acid
Recommended Storage Store at 2 to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.