Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diluted Hydrochloric Acid NF/Dilute Hydrochloric Acid EP, Ricca Chemical
SDP

Catalog No. 371816 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
371816 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 371816 Supplier Ricca Chemical Company Supplier No. 371816
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid
PubChem CID 313
ChEBI CHEBI:23116
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH <1
Appearance Clear colorless to slight greenish yellow
Packaging Amber Glass Bottle
Quantity 500 mL
Physical Form Liquid
Percent Purity 22.6% V/V
Grade EP/NF
Chemical Name or Material Hydrochloric Acid
Recommended Storage Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.