Learn More
Specifications
Specifications
| Packaging | Glass Bottle |
| Quantity | 500 mL |
| IUPAC Name | hydrogen chloride |
| Formula Weight | 36.46 |
| Percent Purity | 36.5% to 38.0% |
| Grade | Not Available |
| Chemical Name or Material | Hydrochloric Acid, (HCl 36.5%-38.0%) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.