Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid NF/FCC/ACS, Macron Fine Chemicals™

Catalog No. MK26126X6 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
10 lb.
4 x 2.5 L
Packaging:
Glass
Glass Bottle
Poly
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-2612-6X6 2.5 L Glass Bottle
MK-2612-500 500 mL Glass Bottle
MK-2612-10 10 lb. Poly
MK-2612212X4 4 x 2.5 L Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MK-2612-6X6 Supplier Avantor Supplier No. 261246
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Packaging Glass Bottle
Quantity 2.5 L
IUPAC Name chlorane
Formula Weight 36.46
Grade ACS/FCC/NF
Chemical Name or Material Hydrochloric Acid, (HCl 36.5%-38.0%)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.