Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, Reagent, ACS, 36.5-38%, Spectrum™ Chemical
SDP

Catalog No. 18603211 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
6 x 500 mL
5 gal.
4 x 4 L
6 x 2.5 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Bottle
Poly Drum
Poly-Coated Glass Bottle
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-211 5 gal. Poly Drum
18-603-201 100 mL Amber Glass Bottle
18-603-204 500 mL Amber Glass Bottle
18-603-207 2.5 L Glass Bottle
18-603-202 2.5 L Poly-Coated Glass Bottle
18-603-203 6 x 500 mL Amber Glass Bottle
18-603-205 4 L Poly Bottle
18-603-209 6 x 2.5 L Glass Bottle
18-603-210 6 x 2.5 L Poly-Coated Glass Bottle
18-603-206 4 x 4 L Poly Bottle
18-603-208 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 18-603-211 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H10355GL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H1035, 7647-01-0, HCl

Spectrum™ Chemical Hydrochloric Acid, Reagent, ACS , is a colorless and clear, highly corrosive, highly pungent solution of hydrogen chloride in water. As an monoprotic acid and one of the six common strong mineral acids, it is the least likely to be subject to an oxidation-reduction reaction. The fact that it is available as a pure reagent make it an excellent acidifying reagent. Hydrochloric acid is also frequently used during chemical analysis to prepare samples for analysis. It reacts with basic compounds, turning them into dissolved chlorides that can then undergo analysis. As an ACS grade Reagent, Spectrum Chemical manufactured Hydrochloric Acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

CAS Max % 100%
Heavy Metals (as Pb) 1 ppm
Packaging Poly Drum
Residue after Ignition 5 ppm
Quantity 5 gal.
MDL Number MFCD00011324 MFCD00792839
UN Number UN1789
Percent Purity 36.5 to 38%
Grade ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.