Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid Solution, 0.1 mol/L (0.1 N), Supelco

Catalog No. 11100249 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-249 1 Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-249 Supplier MilliporeSigma Supelco Supplier No. 1.09973.0050
Add to Cart
Add to Cart
  • Hydrochloric acid solution is utilized in the preparation of volumetric solutions for titration through dilution.
  • The resulting diluted solution is employed for titrating against potassium hydroxide (KOH) in acetonitrile-water mixtures, which are essential for the potentiometric pKa determination of piroxicam and tenoxicam.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrochloric Acid Solution
CAS 7647-01-0
Concentration 0.1 mol/L (0.1 N)
Quantity 1 Bottle
Molecular Weight (g/mol) 36.46
Percent Purity Unknown
Grade Ungraded
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.