Learn More
Sigma Aldrich Fine Chemicals Biosciences Hydrochloric acid solution
Catalog No. 502899006
Encompass
The Product is 6M SOLUTIONon of hydrochloric acid in water. Hydrochloric acid is a strong mineral acid. It participates in the synthesis of alloxan (hexahydropyrimidine-2 4 5 6-tetrone) and hydrated aluminum chloride.
Catalog No. 50-289-9006 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 8442910X2ML
The Product is 6M SOLUTIONon of hydrochloric acid in water. Hydrochloric acid is a strong mineral acid. It participates in the synthesis of alloxan (hexahydropyrimidine-2 4 5 6-tetrone) and hydrated aluminum chloride.