Learn More
Electron Microscopy Sciences Hydrochloric Acid Solution 0.1N, 1 to 2 100 ML
SDP
Catalog No. 5026625
Encompass_Preferred
HCl F.W. 36.46
HCl used to adjust the pH of buffers and fixative solutions.
pH: 1 to 2
Catalog No. 50-266-25 Supplier Electron Microscopy Sciences Supplier No. 16760
HCl F.W. 36.46
HCl used to adjust the pH of buffers and fixative solutions.
pH: 1 to 2