Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid Solution, Volumetric, 6M HCl (6N), Solstice

Catalog No. 60047420 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-420 1 L Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-420 Supplier Solstice Supplier No. 720331L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Volumetric, 6 M HCl (6N)

Solstice's high-quality titration products cover the complete titration workflow and include:
  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH >0.1 (at 20°C)
Color Colorless
Density ∽1.072g/cu.cm at 20°C
Odor Stinging
Packaging Poly bottle
Vapor Pressure 190mmHg (25°C)
Quantity 1 L
Physical Form Liquid
UN Number UN1789
Solubility Information Completely miscible in water
Formula Weight 36.46g/mol
Grade Not Available
Chemical Name or Material Hydrochloric acid solution, Volumetric, 6M HCl (6N)
Show More Show Less
Hazard Statement P305 + P351 + P338
DOTInformation 8/PGII
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.