Learn More
Hydrochloric acid solution, Volumetric, Reag. Ph. Eur., 0.1 M HCl (0.1N), Solstice
Shop All Solstice Research Chemicals ProductsDescription
Solstice's high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| Packaging | Plastic bottle |
| Vapor Pressure | 190 mmHg (25°C) |
| Quantity | 4 x 5 L |
| UN Number | UN1789 |
| IUPAC Name | hydrogen chloride |
| Formula Weight | 36.46 |
| Grade | Reagent Ph. Eur. |
| Chemical Name or Material | Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N) |
Safety and Handling
| DOTInformation | 8 / PGIII |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.