Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, Trace Metal Also meets ACS specifications, Reagents

Catalog No. 01336832
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
Poly Bottle
Safety Coated Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-336-832 2.5 L Poly Bottle
01-336-831 2.5 L Safety Coated Glass
01-336-830 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-336-832 Supplier Reagents Holdings Llc Supplier No. C51011502.5A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrochloric Acid, Trace Metal Also meets ACS specifications, Trace Metal, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Muriatic Acid, Chlorohydric Acid
IUPAC Name water
SMILES O

Specifications

pH 0
Color Colorless
Boiling Point A constant boiling azeotrope with water containing 20.22% hydrogen chloride boils at 227°F.
Packaging Poly Bottle
Specific Gravity 1.18
Quantity 2.5 L
Physical Form Liquid
Percent Purity 36.5% to 38%
Grade ACS
Chemical Name or Material Hydrochloric Acid, Trace Metal Also meets ACS specifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.