Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric Acid, TraceGrade, 34-37%, Spectrum™ Chemical
SDP

Catalog No. 18603215 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
6 x 500 mL
4 x 2.5 L
Packaging:
Amber Glass Bottle
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-215 4 x 2.5 L Poly Bottle
18-603-212 500 mL Amber Glass Bottle
18-603-214 2.5 L Poly Bottle
18-603-213 6 x 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-603-215 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H11222.5LTPLCS4
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H1122, 7647-01-0, HCl

Spectrum™ Chemical Hydrochloric Acid, TraceGrade is a strong, highly corrosive mineral acid with extremely low metal content (over 50 with levels less than 1 PPB). This grade is primarily used as an analytical reagent in elemental analysis determinations using ICP and AA techniques, digestion of water samples and as a doping agent for organic semiconductors.

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

CAS Max % 70%, 30%
Packaging Poly Bottle
Quantity 4 x 2.5 L
UN Number UN1789
Percent Purity 34 to 37%
Grade High Purity Solvent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.