Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrochloric acid, TraceSELECT™ Ultra, Solstice

Catalog No. 60047781 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-047-781 500 mL
600-29-71 250 mL
600-29-70 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-047-781 Supplier Solstice Supplier No. 96208500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TraceSELECT™ Ultra, for ultratrace analysis, 32-35%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Yellow
Density 1.2g/mL at 25°C, 1.15g/mL at 20°C
Vapor Pressure 3.23 psi (21.1°C), 7.93 psi (37.7°C)
Quantity 500 mL
UN Number UN1789
Formula Weight 36.46
Percent Purity 32.0 to 35.0% (T)
Grade TraceSELECT™ Ultra
Chemical Name or Material Hydrochloric acid
Hazard Statement P261-P280-P305 + P351 + P338-P310
DOTInformation 8 / PGII
RTECSNumber 231-595-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.