Learn More
Hach Company Hydrochloric Acid Standard Solution, 2.5 N, 100 mL MDB
Catalog No. 501994557 Shop All Hach Company Products
Encompass
Acid solution for sulfite and chlorine determination, and estimation of lime in soils. 100 mL Marked Dropping Bottle
Catalog No. 50-199-4557 Supplier Hach Company Supplier No. 141832
Acid solution for sulfite and chlorine determination, and estimation of lime in soils. 100 mL Marked Dropping Bottle