Learn More
Apexbio Technology LLC Hydrochlorothiazide 58-93-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063765
Encompass_Preferred
Hydrochlorothiazide (CAS 58-93-5) is a small molecule belonging to the thiazide class of diuretics commonly employed in biomedical research for its actions on renal physiology It targets the sodium-chloride symporter in the distal convoluted tubule of the nephron inhibiting sodium and chloride reabsorption This inhibition elevates urinary excretion of sodium chloride and water resulting in increased urine output and reduced blood volume Researchers frequently utilize hydrochlorothiazide in studies focused on fluid homeostasis hypertension management and kidney function regulation
Catalog No. 50-000-63765 Supplier Apexbio Technology LLC Supplier No. B19505.1
Hydrochlorothiazide (CAS 58-93-5) is a small molecule belonging to the thiazide class of diuretics commonly employed in biomedical research for its actions on renal physiology It targets the sodium-chloride symporter in the distal convoluted tubule of the nephron inhibiting sodium and chloride reabsorption This inhibition elevates urinary excretion of sodium chloride and water resulting in increased urine output and reduced blood volume Researchers frequently utilize hydrochlorothiazide in studies focused on fluid homeostasis hypertension management and kidney function regulation