Learn More
Medchemexpress LLC Hydrochlorothiazide (Standard) | 58-93-5 | 99.5% | 100 UG
SDP
Catalog No. 5000405868
Encompass_Preferred
Hydrochlorothiazide (HCTZ) is an orally active diuretic agent of the thiazide class. It inhibits the transforming TGF-β/Smad signaling pathway. It also has direct vascular relaxant effects by opening the calcium-activated potassium (KCA) channel.
  • Improves cardiac function
  • Reduces fibrosis
  • Has an antihypertensive effect
  • Reduces blood volume by acting on the kidneys to reduce sodium (Na) reabsorption in the distal convoluted tubule
  • Acts on an electroneutral Na+-Cl co-transporter by competing for the chloride site on the transporter, which induces natriuresis and concomitant water loss
  • Increases the reabsorption of calcium
  • Believed to lower peripheral vascular resistance through other mechanisms
Catalog No. 50-004-05868 Supplier Medchemexpress LLC Supplier No. HYB0252S2100UG
Hydrochlorothiazide (HCTZ) is an orally active diuretic agent of the thiazide class. It inhibits the transforming TGF-β/Smad signaling pathway. It also has direct vascular relaxant effects by opening the calcium-activated potassium (KCA) channel.
  • Improves cardiac function
  • Reduces fibrosis
  • Has an antihypertensive effect
  • Reduces blood volume by acting on the kidneys to reduce sodium (Na) reabsorption in the distal convoluted tubule
  • Acts on an electroneutral Na+-Cl co-transporter by competing for the chloride site on the transporter, which induces natriuresis and concomitant water loss
  • Increases the reabsorption of calcium
  • Believed to lower peripheral vascular resistance through other mechanisms