Learn More
Medchemexpress LLC Hydrochlorothiazide (standard) | 58-93-5 | 99.6% | 50 MG
SDP
Catalog No. 5000405412
Encompass_Preferred
Hydrochlorothiazide (Standard) is the analytical standard of Hydrochlorothiazide, intended for research and analytical applications. It is an orally active diuretic agent of the thiazide class that inhibits the transforming TGF-β/Smad signaling pathway. It also has direct vascular relaxant effects by opening the calcium-activated potassium (KCA) channel, improving cardiac function, reducing fibrosis, and exhibiting an antihypertensive effect. This compound is an analytical standard supplied for assay.
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-05412 Supplier Medchemexpress LLC Supplier No. HYB0252R50MG
Hydrochlorothiazide (Standard) is the analytical standard of Hydrochlorothiazide, intended for research and analytical applications. It is an orally active diuretic agent of the thiazide class that inhibits the transforming TGF-β/Smad signaling pathway. It also has direct vascular relaxant effects by opening the calcium-activated potassium (KCA) channel, improving cardiac function, reducing fibrosis, and exhibiting an antihypertensive effect. This compound is an analytical standard supplied for assay.
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications