Learn More
Selleck Chemical LLC Hydrocinnamic acid
SDP
Catalog No. 5000003711
Encompass_Preferred
Hydrocinnamic acid (3-phenylpropionic acid Benzylacetic acid) is a substrate of the enzyme oxidoreductases in the pathway phenylalanine metabolism (KEGG) It is an antifungal agent and a metabolite
Catalog No. 50-000-03711 Supplier Selleck Chemical LLC Supplier No. S625425MG
Hydrocinnamic acid (3-phenylpropionic acid Benzylacetic acid) is a substrate of the enzyme oxidoreductases in the pathway phenylalanine metabolism (KEGG) It is an antifungal agent and a metabolite