Learn More
Medchemexpress LLC Hydrocinnamic acid-d9 | 93131-15-8 | 99.7% | 159.23 g/mol | C9HD9O2 | 25mg
SDP
Catalog No. 5000267092
Encompass_Preferred
Hydrocinnamic acid-d9 is the deuterium labeled Hydrocinnamic acid[1] Hydrocinnamic acid is the major rhizospheric compound with known growth regulatory activitie[2]s
Catalog No. 50-002-67092 Supplier Medchemexpress LLC Supplier No. HYY1088S225MG
Hydrocinnamic acid-d9 is the deuterium labeled Hydrocinnamic acid[1] Hydrocinnamic acid is the major rhizospheric compound with known growth regulatory activitie[2]s