Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aobchem Hydrocinnamoyl chloride | 95% | CAS 645-45-4 | C9H9ClO | 100g

Catalog No. 5001021215
Encompass_Preferred

Hydrocinnamoyl chloride | 95% | CAS 645-45-4 | C9H9ClO | 100g

Catalog No. 50-010-21215 Supplier Aobchem Supplier No. A406110100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrocinnamoyl chloride | 95% | CAS 645-45-4 | C9H9ClO | 100g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.