Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrocinnamoyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
645-45-4
C9H9ClO
168.62
MFCD00000748
MFEILWXBDBCWKF-UHFFFAOYSA-N
3-phenylpropionyl chloride,hydrocinnamoyl chloride,benzenepropanoyl chloride,dihydrocinnamoyl chloride,hydrocinnamyl chloride,propionyl chloride, 3-phenyl,hydrocinnamoylchloride,beta-phenylpropionyl chloride,3-phenylpropionic acid chloride,beta-phenylpropanoyl chloride
64801
3-phenylpropanoyl chloride
C1=CC=C(C=C1)CCC(=O)Cl
645-45-4
C9H9ClO
168.62
MFCD00000748
MFEILWXBDBCWKF-UHFFFAOYSA-N
3-phenylpropionyl chloride,hydrocinnamoyl chloride,benzenepropanoyl chloride,dihydrocinnamoyl chloride,hydrocinnamyl chloride,propionyl chloride, 3-phenyl,hydrocinnamoylchloride,beta-phenylpropionyl chloride,3-phenylpropionic acid chloride,beta-phenylpropanoyl chloride
64801
3-phenylpropanoyl chloride
C1=CC=C(C=C1)CCC(=O)Cl
Melting Point -6°C
Specific Gravity 1.137
Boiling Point 107°C (11.0mmHg)
Flash Point 108°C
Density 1.1370g/mL
Refractive Index 1.5260 to 1.528
Color Colorless to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H5(CH2)2COCl
Beilstein 09, 511
Solubility Information Solubility in water: decomposes in water. Other solubilities: soluble in ether
Formula Weight 168.62
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Hydrocinnamoyl chloride, 98%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-443-6

RTECSNumber : MW5652395

TSCA : TSCA