Learn More
Andwin Scientific HYDROCLRC ACD 6.0N 1L
SDP
Catalog No. 5089990341
Encompass_Preferred
The Hydrochloric Acid solution has CAS number of 7647-01-0. It has a concentration of 6.0 M. This chemical is in a form of clear, colorless liquid that has a highly pungent odor. It is one of the six common strong mineral acids it is the least likely to be subject to an oxidation-reduction reaction. This product reacts with basic compounds. It is packaged and sold in a 1-liter amber glass bottle.
Catalog No. 50-899-90341 Supplier Andwin Scientific Supplier No. LC153702
The Hydrochloric Acid solution has CAS number of 7647-01-0. It has a concentration of 6.0 M. This chemical is in a form of clear, colorless liquid that has a highly pungent odor. It is one of the six common strong mineral acids it is the least likely to be subject to an oxidation-reduction reaction. This product reacts with basic compounds. It is packaged and sold in a 1-liter amber glass bottle.