Learn More
Cayman Chemical Hydrocortisone Aceponate, Purity: ≥98%, CAS: 74050-20-7, 100 mg
SDP
Catalog No. 502909426
Encompass_Preferred
A corticosteroid; reduces lesion size and pruritis in cats with allergic dermatitis when applied topically at 0.0584%; induces remission and increases time to relapse in dogs with spontaneous atopic dermatitis
Catalog No. 50-290-9426 Supplier Cayman Chemical Supplier No. 35359100 MG
A corticosteroid; reduces lesion size and pruritis in cats with allergic dermatitis when applied topically at 0.0584%; induces remission and increases time to relapse in dogs with spontaneous atopic dermatitis