Learn More
Cayman Chemical Hydrocortisone Acetate, Purity: ≥98%, CAS: 50-03-3, 25 g
SDP
Catalog No. 502989162
Encompass_Preferred
A synthetic corticosteroid; topical administration reduces corneal haze and hydroxyproline levels without reducing the stromal wound healing response in a rabbit model of photorefractive keratectomy at 0.1% v/v; enhances and reduces cartilage degradation in a mouse model of air pouch cartilage implantation when injected into the air pouch cavity or into the air pouch lining tissue, respectively
Catalog No. 50-298-9162 Supplier Cayman Chemical Supplier No. 2939025 G
A synthetic corticosteroid; topical administration reduces corneal haze and hydroxyproline levels without reducing the stromal wound healing response in a rabbit model of photorefractive keratectomy at 0.1% v/v; enhances and reduces cartilage degradation in a mouse model of air pouch cartilage implantation when injected into the air pouch cavity or into the air pouch lining tissue, respectively