Learn More
Cayman Chemical Hydrocortisone-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 73565-87-4, 1 mg
SDP
Catalog No. 502935448
Encompass_Preferred
An internal standard for the quantification of hydrocortisone/cortisol by GC- or LC-MS
Catalog No. 50-293-5448 Supplier Cayman Chemical Supplier No. 265001 MG
An internal standard for the quantification of hydrocortisone/cortisol by GC- or LC-MS