Learn More
Cayman Chemical HydrocortIson-d4 500ug
SDP
Catalog No. 502933618
Encompass_Preferred
An internal standard for the quantification of hydrocortisone/cortisol by GC- or LC-MS
Catalog No. 50-293-3618 Supplier Cayman Chemical Supplier No. 26500500 UG
An internal standard for the quantification of hydrocortisone/cortisol by GC- or LC-MS