Learn More
Medchemexpress LLC Hydrocortisone 17-butyrate | 13609-67-1 | 99.9% | 1 ML
Supplier: Medchemexpress LLC HYB098310MM/1ML
Hydrocortisone 17-butyrate is a non-fluorinated topical corticosteroid. It is used for psoriasis, eczema, and other inflammatory dermatoses. This compound has been shown to inhibit granuloma formation in rats and can induce vasoconstriction.
- Non-fluorinated topical corticosteroid
- Used for psoriasis, eczema, and other inflammatory dermatoses
- Inhibits granuloma formation in rats
- Can induce vasoconstriction