Learn More
Medchemexpress LLC Hydrocortisone 17-butyrate | 13609-67-1 | 99.94% | 100 MG
SDP
Catalog No. 5000408273
Encompass_Preferred
Hydrocortisone 17-butyrate is a non-fluorinated topical corticosteroid for use in psoriasis, eczema, and other inflammatory dermatoses. It inhibits granuloma formation in rats and can induce vasoconstriction.
  • Non-fluorinated topical corticosteroid
  • Used for psoriasis, eczema, and inflammatory dermatoses
  • Inhibits granuloma formation
  • Induces vasoconstriction
Catalog No. 50-004-08273 Supplier Medchemexpress LLC Supplier No. HYB0983100MG
Hydrocortisone 17-butyrate is a non-fluorinated topical corticosteroid for use in psoriasis, eczema, and other inflammatory dermatoses. It inhibits granuloma formation in rats and can induce vasoconstriction.
  • Non-fluorinated topical corticosteroid
  • Used for psoriasis, eczema, and inflammatory dermatoses
  • Inhibits granuloma formation
  • Induces vasoconstriction