Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydrocortisone 17-valerate | 57524-89-7 | MFCD00083365 | 98.0% | 446.58 g/mol | C26H38O6 | 10 MG
SDP

Catalog No. 5000298217
Encompass_Preferred

Hydrocortisone 17-valerate is a corticosteroid ester used as an analytical standard and research reagent with anti-inflammatory activity. It is provided for laboratory research applications and intended for research use only.

  • Analytical standard for hydrocortisone derivatives.
  • Purity 98.0% for reliable analytical results.
  • Molecular weight 446.58 g/mol; chemical formula C26H38O6.
  • Available in small pack sizes suitable for analytical and method development work.
  • Suitable for in vitro research and analytical assays.
  • CAS number 57524-89-7 for unambiguous identification.

Catalog No. 50-002-98217 Supplier Medchemexpress LLC Supplier No. HYB126510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrocortisone 17-valerate is a corticosteroid ester used as an analytical standard and research reagent with anti-inflammatory activity. It is provided for laboratory research applications and intended for research use only.

  • Analytical standard for hydrocortisone derivatives.
  • Purity 98.0% for reliable analytical results.
  • Molecular weight 446.58 g/mol; chemical formula C26H38O6.
  • Available in small pack sizes suitable for analytical and method development work.
  • Suitable for in vitro research and analytical assays.
  • CAS number 57524-89-7 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.