Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrocortisone-21-Acetate, MP Biomedicals™

Catalog No. ICN10199505 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10199505 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10199505 Supplier MP Biomedicals, Inc Supplier No. 0210199505

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50-03-3
Molecular Formula C23H32O6
Molecular Weight (g/mol) 404.50
MDL Number MFCD00037714
InChI Key ALEXXDVDDISNDU-JZYPGELDSA-N
Synonym hydrocortisone acetate, cortell, cortisol 21-acetate, cortisol acetate, cortacream, hydrocortisat, abbocort, bambicort, biocortar, chemysone
PubChem CID 5744
ChEBI CHEBI:17609
SMILES CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C

Specifications

Color White
Quantity 5 g
IUPAC Name 2-[(1R,3aS,3bS,9aR,9bS,10S,11aS)-1,10-dihydroxy-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
Formula Weight 404.5
Physical Form Solid
Chemical Name or Material Hydrocortisone-21-Acetate
EINECSNumber 200-004-4
storageNote1 Store desiccated, protect from light
Recommended Storage Store at Room Temperature (15° to 30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.