Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Hydrocortisone-21-Acetate, MP Biomedicals™

Catalog No. MP210199525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210199525 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210199525 Supplier MP Biomedicals™ Supplier No. 0210199525

missing translation for 'validMainframeMsg'

Inhibits most inflammatory and immune responses of allergic, bacterial, traumatic or anaphylactic origin. Hydrocortisone-21-Acetate, MP Biomedicals is an immunosuppressant drug that acts by inhibiting migration of leukocytes and phagocytes and decreasing

  • Classified as a corticosteroid (e.g. betamethasone, dexamethasone, fluorometholone, hydrocortisone acetate, loteprednol etabonate, prednisolone, rimexolone, triamcinolone)
  • They are sometimes combined with an antibiotic drug (e.g. betamethasone combined with neomycin or sulfacetamide, dexamethasone combined with neomycin or polymyxin B)
  • Primary functions are to increase blood sugar through gluconeogenesis and to suppresses inflammatory and immune responses
TRUSTED_SUSTAINABILITY

Specifications

Color White
Content And Storage Store at Room Temperature; desiccate. Protect from light.
Form Powder
Quantity 25 g
Regulatory Status RUO
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.