Learn More
Medchemexpress LLC Hydrocortisone 21-hemisuccinate sodium | 125-04-2 | 99.7% | 100 MG
SDP
Catalog No. 5000412124
Encompass_Preferred
Hydrocortisone hemisuccinate sodium is an orally active physiological glucocorticoid that inhibits proinflammatory cytokine activity, with IC50s of 6.7 μM for IL-6 and 21.4 μM for IL-3. This compound can be used for the research of ulcerative colitis (UC).
  • Orally active physiological glucocorticoid
  • Inhibits proinflammatory cytokine activity (IL-6 and IL-3)
  • Can be used for research of ulcerative colitis (UC)
  • Reduces weight loss and increases food intake in mice at 30 mg/kg dosage
  • Significantly decreased disease activity index (DAI) scores and myeloperoxidase (MPO) activity in a rat model of colitis
Catalog No. 50-004-12124 Supplier Medchemexpress LLC Supplier No. HYB1402B100MG
Hydrocortisone hemisuccinate sodium is an orally active physiological glucocorticoid that inhibits proinflammatory cytokine activity, with IC50s of 6.7 μM for IL-6 and 21.4 μM for IL-3. This compound can be used for the research of ulcerative colitis (UC).
  • Orally active physiological glucocorticoid
  • Inhibits proinflammatory cytokine activity (IL-6 and IL-3)
  • Can be used for research of ulcerative colitis (UC)
  • Reduces weight loss and increases food intake in mice at 30 mg/kg dosage
  • Significantly decreased disease activity index (DAI) scores and myeloperoxidase (MPO) activity in a rat model of colitis