Learn More
Apexbio Technology LLC Hydrocortisone(Synonyms: Cortisol, 17-Hydroxycorticosterone, Kendall's Compound F), 5g, CAS: 50-23-7.
SDP
Catalog No. 5000060096
Encompass_Preferred
Hydrocortisone (CAS 50-23-7) is an endogenous glucocorticoid primarily synthesized and secreted by the adrenal cortex It exerts biological effects by binding glucocorticoid receptors altering gene expression involved in metabolic regulation immune response modulation and anti-inflammatory pathways Widely utilized in biomedical research hydrocortisone serves as a standard reference agent for investigating glucocorticoid receptor signaling inflammation models and stress response mechanisms
Catalog No. 50-000-60096 Supplier Apexbio Technology LLC Supplier No. B19515000
Add to Cart
Add to Cart
Hydrocortisone (CAS 50-23-7) is an endogenous glucocorticoid primarily synthesized and secreted by the adrenal cortex It exerts biological effects by binding glucocorticoid receptors altering gene expression involved in metabolic regulation immune response modulation and anti-inflammatory pathways Widely utilized in biomedical research hydrocortisone serves as a standard reference agent for investigating glucocorticoid receptor signaling inflammation models and stress response mechanisms