Learn More
Medchemexpress LLC HY-N0583 5g , Hydrocortisone Cortisol CAS:50-23-7 Purity:98%
SDP
Catalog No. 501492256
Encompass_Preferred
Medchemexpress, HY-N0583 5g Hydrocortisone Cortisol CAS:50-23-7 Formula:C21H30O5 Purity:98% Hydrocortisone is a steroid hormone or glucocorticoid secreted by the adrenal cortex. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2256 Supplier Medchemexpress LLC Supplier No. HYN05835G
Medchemexpress, HY-N0583 5g Hydrocortisone Cortisol CAS:50-23-7 Formula:C21H30O5 Purity:98% Hydrocortisone is a steroid hormone or glucocorticoid secreted by the adrenal cortex. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.