Learn More
Sigma Aldrich Fine Chemicals Biosciences Hydrocortisone, 50-23-7, MFCD00011654, 1g
Catalog No. NC1631125
Encompass
Empirical Formula (Hill Notation) C21H30O5, ≥98% (HPLC), Molecular Weight 362.46, mp 211-214 °C (lit.), Synonym: 11β,17α,21-Trihydroxypregn-4-ene-3,20-dione, 17-Hydroxycorticosterone, 4-Pregnene-11β,17α,21-triol-3,20-dione, Cortisol, Kendall’s compound F, Reichstein’s substance M.
Catalog No. NC1631125 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H40011G
Empirical Formula (Hill Notation) C21H30O5, ≥98% (HPLC), Molecular Weight 362.46, mp 211-214 °C (lit.), Synonym: 11β,17α,21-Trihydroxypregn-4-ene-3,20-dione, 17-Hydroxycorticosterone, 4-Pregnene-11β,17α,21-triol-3,20-dione, Cortisol, Kendall’s compound F, Reichstein’s substance M.