Learn More
Medchemexpress LLC Hydrocortisone acepo 10mg | 74050-20-7 | 460.56 | C26H36O7 | 10 MG
SDP
Catalog No. 5000226160
Encompass_Preferred
Hydrocortisone aceponate (hydrocortisone 17-propionate 21-acetate) analytical standard supplied as a 10 mg package with 97% purity for research and analytical applications. It is used as a topical glucocorticoid reference standard in chromatographic and mass-spectrometric methods and in dermatoses research.
  • Analytical reference standard for HPLC, GC, and MS methods.
  • High purity (97%) suitable for quantitative and qualitative analysis.
  • Supplied as a 10 mg package for small-scale laboratory use.
  • Molecular formula C26H36O7 and molecular weight 460.56 g/mol for identification and calculations.
  • Intended for research use only; not for human or veterinary administration.
Catalog No. 50-002-26160 Supplier Medchemexpress LLC Supplier No. HY116691R10MG
Hydrocortisone aceponate (hydrocortisone 17-propionate 21-acetate) analytical standard supplied as a 10 mg package with 97% purity for research and analytical applications. It is used as a topical glucocorticoid reference standard in chromatographic and mass-spectrometric methods and in dermatoses research.
  • Analytical reference standard for HPLC, GC, and MS methods.
  • High purity (97%) suitable for quantitative and qualitative analysis.
  • Supplied as a 10 mg package for small-scale laboratory use.
  • Molecular formula C26H36O7 and molecular weight 460.56 g/mol for identification and calculations.
  • Intended for research use only; not for human or veterinary administration.