Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydrocortisone acepo 10mg | 74050-20-7 | 460.56 g/mol | C26H36O7 | 10 MG
SDP

Catalog No. 5000226152
Encompass_Preferred

Hydrocortisone aceponate (hydrocortisone 17-propionate 21-acetate) is a potent topical glucocorticoid supplied for research use in studies of dermatoses and glucocorticoid receptor biology.

  • Potent topical glucocorticoid for dermatoses research.
  • White to off-white powder appearance.
  • Purity approximately 99.4% as stated by supplier.
  • Chemical formula C26H36O7; molecular weight 460.56 g/mol.
  • Available as solids (5 mg, 10 mg, 25 mg, 50 mg) and as a 10 mM solution in DMSO (1 mL).
  • Soluble in DMSO (~100 mg/mL); ultrasonic treatment recommended.
  • Storage: powder -20°C (stable ~3 years) or 4°C (stable ~2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-26152 Supplier Medchemexpress LLC Supplier No. HY11669110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Hydrocortisone aceponate (hydrocortisone 17-propionate 21-acetate) is a potent topical glucocorticoid supplied for research use in studies of dermatoses and glucocorticoid receptor biology.

  • Potent topical glucocorticoid for dermatoses research.
  • White to off-white powder appearance.
  • Purity approximately 99.4% as stated by supplier.
  • Chemical formula C26H36O7; molecular weight 460.56 g/mol.
  • Available as solids (5 mg, 10 mg, 25 mg, 50 mg) and as a 10 mM solution in DMSO (1 mL).
  • Soluble in DMSO (~100 mg/mL); ultrasonic treatment recommended.
  • Storage: powder -20°C (stable ~3 years) or 4°C (stable ~2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.