Learn More
Medchemexpress LLC Hydrocortisone acepo 10mM 1mL | 74050-20-7 | 460.56 g/mol | C26H36O7 | 1 ML
SDP
Catalog No. 5000227161
Encompass_Preferred
Hydrocortisone aceponate (hydrocortisone 17-propionate 21-acetate) is a synthetic topical glucocorticoid supplied as an analytical standard for research applications. It is offered as solid material and as a ready-to-use 10 mM solution in DMSO (1 mL). The compound is used in dermatoses and glucocorticoid receptor studies; supplier guidance lists purity and storage conditions.
Catalog No. 50-002-27161 Supplier Medchemexpress LLC Supplier No. HY11669110MM 1ML
Hydrocortisone aceponate (hydrocortisone 17-propionate 21-acetate) is a synthetic topical glucocorticoid supplied as an analytical standard for research applications. It is offered as solid material and as a ready-to-use 10 mM solution in DMSO (1 mL). The compound is used in dermatoses and glucocorticoid receptor studies; supplier guidance lists purity and storage conditions.