Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydrocortisone acepo 25mg | 74050-20-7 | 460.56 g/mol | C26H36O7 | 25 MG
SDP

Catalog No. 5000308020
Encompass_Preferred

Hydrocortisone aceponate is supplied as an analytical standard for laboratory research into topical glucocorticoid activity. It is provided as a small solid pack appropriate for in vitro and analytical assays, with reported high purity and defined molecular properties.

  • Purity listed as 97% for reliable analytical use.
  • Molecular formula C26H36O7 and molecular weight 460.56 g/mol for accurate calculations.
  • CAS number 74050-20-7 for unambiguous compound identification.
  • Available in small pack sizes suitable for method development and standards preparation.
  • Intended application: analytical and research use for dermal glucocorticoid studies.

Catalog No. 50-003-08020 Supplier Medchemexpress LLC Supplier No. HY116691R25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Hydrocortisone aceponate is supplied as an analytical standard for laboratory research into topical glucocorticoid activity. It is provided as a small solid pack appropriate for in vitro and analytical assays, with reported high purity and defined molecular properties.

  • Purity listed as 97% for reliable analytical use.
  • Molecular formula C26H36O7 and molecular weight 460.56 g/mol for accurate calculations.
  • CAS number 74050-20-7 for unambiguous compound identification.
  • Available in small pack sizes suitable for method development and standards preparation.
  • Intended application: analytical and research use for dermal glucocorticoid studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.