Learn More
Medchemexpress LLC Hydrocortisone acepo 25mg | 74050-20-7 | 460.6 g/mol | C26H36O7 | 25 MG
SDP
Catalog No. 5000227162
Encompass_Preferred
Hydrocortisone aceponate (Hydrocortisone 17-propionate 21-acetate, CAS 74050-20-7) is a topical glucocorticoid research compound used to study anti-inflammatory and antipruritic effects in dermatological and veterinary settings. It is supplied for formulation, pharmacology, and analytical research, typically as a high-purity solid suitable for analytical standards and small-scale formulation work.
  • Topical glucocorticoid with anti-inflammatory and antipruritic activity.
  • Suitable for dermatological and veterinary research applications.
  • Supplied as a research-grade solid, typically ≥98% purity.
  • Available in small analytical/package sizes, e.g., 25 MG, for formulation and assay work.
  • Soluble in common organic solvents such as chloroform and methanol for analytical preparation.
Catalog No. 50-002-27162 Supplier Medchemexpress LLC Supplier No. HY11669125MG
Hydrocortisone aceponate (Hydrocortisone 17-propionate 21-acetate, CAS 74050-20-7) is a topical glucocorticoid research compound used to study anti-inflammatory and antipruritic effects in dermatological and veterinary settings. It is supplied for formulation, pharmacology, and analytical research, typically as a high-purity solid suitable for analytical standards and small-scale formulation work.
  • Topical glucocorticoid with anti-inflammatory and antipruritic activity.
  • Suitable for dermatological and veterinary research applications.
  • Supplied as a research-grade solid, typically ≥98% purity.
  • Available in small analytical/package sizes, e.g., 25 MG, for formulation and assay work.
  • Soluble in common organic solvents such as chloroform and methanol for analytical preparation.