Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydrocortisone acepo 50mg | 74050-20-7 | 460.56 | C26H36O7 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY11669150MG

Encompass_Preferred

Hydrocortisone aceponate (hydrocortisone 17-propionate 21-acetate) is a potent topical glucocorticoid supplied as a research-grade chemical standard for dermatological and pharmacological studies. The compound is supplied as a solid with high purity and is suitable for analytical and experimental applications.

  • Potent topical glucocorticoid for dermatoses research.
  • High purity (99.43%).
  • Solid, white to off-white appearance.
  • Molecular formula C26H36O7 and molecular weight 460.56.
  • Available in multiple pack sizes, including 50 mg.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-30021


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.