Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydrocortisone acepo 50mg | 74050-20-7 | 460.56 g/mol | C26H36O7 | 50 MG
SDP

Catalog No. 5000227525
Encompass_Preferred

Hydrocortisone aceponate (hydrocortisone 17-propionate 21-acetate) is a topical glucocorticoid supplied as a research/analytical standard for laboratory use. It is provided as a solid reference material (CAS 74050-20-7) for in vitro and analytical studies and is not intended for therapeutic use.

  • Topical glucocorticoid for dermatoses research.
  • Provided as a research/analytical standard for laboratory applications.
  • CAS number 74050-20-7 and chemical formula C26H36O7.
  • Molecular weight ~460.56 g/mol.
  • Purity reported by vendors; consult the certificate of analysis before use.
  • Available in multiple pack sizes, including 50 mg.

Catalog No. 50-002-27525 Supplier Medchemexpress LLC Supplier No. HY116691R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Hydrocortisone aceponate (hydrocortisone 17-propionate 21-acetate) is a topical glucocorticoid supplied as a research/analytical standard for laboratory use. It is provided as a solid reference material (CAS 74050-20-7) for in vitro and analytical studies and is not intended for therapeutic use.

  • Topical glucocorticoid for dermatoses research.
  • Provided as a research/analytical standard for laboratory applications.
  • CAS number 74050-20-7 and chemical formula C26H36O7.
  • Molecular weight ~460.56 g/mol.
  • Purity reported by vendors; consult the certificate of analysis before use.
  • Available in multiple pack sizes, including 50 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.