Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydrocortisone acepo 5mg | 74050-20-7 | 460.56 | C26H36O7 | 5 MG
SDP

Catalog No. 5000230022
Encompass_Preferred

Hydrocortisone aceponate (Hydrocortisone 17-propionate 21-acetate) is a potent topical glucocorticoid supplied as a research-grade analytical standard in solid powder and solution forms. It is used for studies of glucocorticoid receptor activity and for research into inflammatory dermatoses. Molecular formula C26H36O7; molecular weight 460.56; CAS 74050-20-7.

  • Potent topical glucocorticoid for dermatoses research.
  • Provided as powder (solid) and as 10 mM solution in DMSO.
  • Intended for research and analytical applications (research use only).
  • Target: glucocorticoid receptor; pathway: immunology/inflammation.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Analytical documentation available: COA, HNMR, LCMS.

Catalog No. 50-002-30022 Supplier Medchemexpress LLC Supplier No. HY1166915MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrocortisone aceponate (Hydrocortisone 17-propionate 21-acetate) is a potent topical glucocorticoid supplied as a research-grade analytical standard in solid powder and solution forms. It is used for studies of glucocorticoid receptor activity and for research into inflammatory dermatoses. Molecular formula C26H36O7; molecular weight 460.56; CAS 74050-20-7.

  • Potent topical glucocorticoid for dermatoses research.
  • Provided as powder (solid) and as 10 mM solution in DMSO.
  • Intended for research and analytical applications (research use only).
  • Target: glucocorticoid receptor; pathway: immunology/inflammation.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Analytical documentation available: COA, HNMR, LCMS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.