Learn More
Medchemexpress LLC Hydrocortisone acepo 5mg | 74050-20-7 | 460.56 g·mol⁻¹ | C26H36O7 | 5 MG
SDP
Catalog No. 5000230023
Encompass_Preferred
Hydrocortisone aceponate is supplied as an analytical standard of hydrocortisone 17-propionate 21-acetate for research and analytical applications, commonly used as a reference in assay development and dermatoses studies.
  • Analytical standard of hydrocortisone aceponate for research and assays.
  • Declared purity suitable for HPLC analysis (97%).
  • Molecular formula C26H36O7 and molecular weight 460.56 g·mol⁻¹.
  • Supplied in a small 5 mg quantity for analytical use.
  • Suitable as a reference standard in topical glucocorticoid research.
Catalog No. 50-002-30023 Supplier Medchemexpress LLC Supplier No. HY116691R5MG
Hydrocortisone aceponate is supplied as an analytical standard of hydrocortisone 17-propionate 21-acetate for research and analytical applications, commonly used as a reference in assay development and dermatoses studies.
  • Analytical standard of hydrocortisone aceponate for research and assays.
  • Declared purity suitable for HPLC analysis (97%).
  • Molecular formula C26H36O7 and molecular weight 460.56 g·mol⁻¹.
  • Supplied in a small 5 mg quantity for analytical use.
  • Suitable as a reference standard in topical glucocorticoid research.