Learn More
TARGETMOL CHEMICALS INC HYDROCORTISONE ACEPONATE 5MG
SDP
Catalog No. 502580337
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Hydrocortisone aceponate (Hydrocortisone 17-propionate 21-acetate) is an effective topical glucocorticoid used in the form of a cream to treat various skin diseases. purity: 99%
Catalog No. 50-258-0337 Supplier TARGETMOL CHEMICALS INC Supplier No. T321145MG
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Hydrocortisone aceponate (Hydrocortisone 17-propionate 21-acetate) is an effective topical glucocorticoid used in the form of a cream to treat various skin diseases. purity: 99%