Learn More
Abcam Hydrocortisone, Adrenal glucocorticoid, 5G
Catalog No. 502706761
Encompass_Preferred
MW 362.5 Da, Purity >99%. Adrenal glucocorticoid. Stimulates gluconeogenesis, activates anxiolytic and anti-inflammatory pathways.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6761 Supplier Abcam Supplier No. AB1412505G
MW 362.5 Da, Purity >99%. Adrenal glucocorticoid. Stimulates gluconeogenesis, activates anxiolytic and anti-inflammatory pathways.
The product is subject to the following: Abcam Restricted Use Statement