Learn More
Medchemexpress LLC Hydrocortisone butep 100mg | 72590-77-3 | 100MG
SDP
Catalog No. 5000242033
Encompass_Preferred
Hydrocortisone buteprate (Hydrocortisone probutate) is a medium potent non-halogenated double-ester of hydrocortisone with a favorable benefit/risk ratio for the treatment of inflammatory skin disorders Hydrocortisone buteprate (Hydrocortisone probutate) is available as a 0 1% cream or ointment formulation[1
Catalog No. 50-002-42033 Supplier Medchemexpress LLC Supplier No. HY106673100MG
Hydrocortisone buteprate (Hydrocortisone probutate) is a medium potent non-halogenated double-ester of hydrocortisone with a favorable benefit/risk ratio for the treatment of inflammatory skin disorders Hydrocortisone buteprate (Hydrocortisone probutate) is available as a 0 1% cream or ointment formulation[1